Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100pbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029036.2 | NP_083312.1 | 3 | 297 | ||
GenBank mRNA | AK015229.1 | - | 3 | 297 | ||
AK030636.1 | - | 3 | 225 | |||
AK046756.1 | - | 2 | 108 | |||
AK139097.1 | - | 3 | 271 | |||
AK146310.1 | - | 3 | 360 | |||
AK169364.1 | - | 3 | 262 | |||
BC038329.1 | - | 3 | 304 | 1548746 |