Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XKR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018053.3 | NP_060523.2 | 3 | 2072 | ||
XM_011541679.2 | XP_011539981.1 | 5 | 2343 | |||
XM_011541680.2 | XP_011539982.1 | 4 | 2235 | |||
GenBank mRNA | AK001169.1 | - | 1 | 739 | ||
AK026024.1 | - | 3 | 1541 | |||
AK054909.1 | - | 1 | 2165 | |||
AK091615.1 | - | 3 | 2072 | |||
AY534246.1 | - | 3 | 1530 | |||
BC003606.1 | - | 1 | 959 | |||
BC013379.2 | - | 3 | 1531 | 4025274 | ||
BC028564.2 | - | 3 | 1531 | 3832072 |
Gene Symbol: RNF216P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124529.1 | - | 2 | 3008 | ||
BC026736.1 | - | 6 | 1509 | |||
BX538059.1 | - | 6 | 1557 |