Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008506.2 | NP_001008506.2 | 15 | 2409 | ||
NM_001160107.1 | NP_001153579.1 | 14 | 2334 | |||
NM_001160108.1 | NP_001153580.1 | 7 | 1603 | |||
NM_029334.2 | NP_083610.2 | 17 | 2802 | |||
GenBank mRNA | AK048046.1 | - | 17 | 2768 | ||
AK050226.1 | - | 17 | 2783 | |||
AK160937.1 | - | 14 | 2320 | |||
BC024856.1 | - | 10 | 1424 | |||
BC034760.1 | - | 17 | 2735 |
Gene Symbol: 3110067C02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK081077.1 | - | 1 | 1969 |