Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppfia3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029741.2 | NP_084017.2 | 11 | 1692 | ||
GenBank mRNA | AB516303.1 | - | 11 | 1474 | ||
AK147645.1 | - | 11 | 1692 | |||
BC058404.1 | - | 9 | 1146 | 6827530 | ||
EU568871.1 | - | 11 | 1602 |