Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029749.2 | NP_084025.2 | 15 | 3329 | ||
GenBank mRNA | AK029419.1 | - | 1 | 939 | ||
AK041514.1 | - | 2 | 2021 | |||
BC137852.1 | - | 14 | 3222 | 8860871 |