Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spopl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165997.1 | NP_001159469.1 | 12 | 2539 | ||
NM_001165998.1 | NP_001159470.1 | 11 | 2403 | |||
NM_029773.2 | NP_084049.2 | 11 | 2442 | |||
GenBank mRNA | AV228319.2 | - | 1 | 306 |