Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spopl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165997.1 | NP_001159469.1 | 5 | 684 | ||
NM_001165998.1 | NP_001159470.1 | 4 | 548 | |||
NM_029773.2 | NP_084049.2 | 5 | 684 | |||
GenBank mRNA | BC111867.1 | - | 5 | 558 | 40056321 | |
CF746115.1 | - | 2 | 137 |