Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052847.2 | NP_443079.1 | 5 | 1010 | ||
XM_017026606.1 | XP_016882095.1 | 5 | 1119 | |||
GenBank mRNA | AK123863.1 | - | 1 | 14 | ||
BC053630.1 | - | 5 | 925 | 6469872 |