Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1293 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000460.1 | NP_001000460.1 | 1 | 574 |
Gene Symbol: LOC100910599 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770723.1 | XP_008768945.1 | 1 | 574 |