Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KBTBD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318716.1 | NP_001305645.1 | 4 | 2482 | ||
NM_001318717.1 | NP_001305646.1 | 4 | 2403 | |||
NM_001318718.1 | NP_001305647.1 | 4 | 2383 | |||
NM_001318719.1 | NP_001305648.1 | 4 | 2361 | |||
NM_001318720.1 | NP_001305649.1 | 4 | 2355 | |||
NM_001318721.1 | NP_001305650.1 | 4 | 2330 | |||
NM_001318722.1 | NP_001305651.1 | 4 | 2351 | |||
NM_001318723.1 | NP_001305652.1 | 4 | 2435 | |||
NM_001318724.1 | NP_001305653.1 | 4 | 2435 | |||
NM_001318725.1 | NP_001305654.1 | 4 | 2338 | |||
NM_016506.6 | NP_057590.3 | 4 | 2439 | |||
NM_018095.5 | NP_060565.4 | 4 | 2334 | |||
XM_017018009.1 | XP_016873498.1 | 4 | 2318 | |||
GenBank mRNA | AK001312.1 | - | 4 | 2288 | ||
AK001749.1 | - | 4 | 2375 | |||
AK091607.1 | - | 3 | 3012 | |||
AK222985.1 | - | 5 | 1475 | |||
AK225062.1 | - | 4 | 2285 | |||
AL713735.1 | - | 3 | 2264 | |||
BC002736.2 | - | 4 | 2376 | 3628290 |