Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RBM23 SIRNA Assays ›
Gene Symbol:
RBM23
Gene Name:
RNA binding motif protein 23
Gene Aliases:
CAPERbeta, PP239, RNPC4
Chromosome Location:
Chr.14: 22900645 - 22919187 on Build GRCh38
UniGene ID:
Hs.4997
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
26266
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RBM23
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077351.1
NP_001070819.1
10
1120
NM_001077352.1
NP_001070820.1
8
1018
NM_001308044.1
NP_001294973.1
8
959
NM_018107.4
NP_060577.3
9
1072
XM_011536890.2
XP_011535192.1
11
1203
XM_011536892.2
XP_011535194.1
10
1154
XM_011536893.2
XP_011535195.1
10
1155
XM_011536894.2
XP_011535196.1
10
1149
XM_011536895.2
XP_011535197.1
9
1125
XM_011536896.2
XP_011535198.1
9
1101
XM_011536897.2
XP_011535199.1
9
1487
XM_011536900.1
XP_011535202.1
9
1063
XM_011536902.1
XP_011535204.1
9
969
XM_011536903.1
XP_011535205.1
7
845
XM_011536904.1
XP_011535206.1
8
902
XM_011536905.1
XP_011535207.1
7
900
XM_011536906.1
XP_011535208.1
6
791
XM_017021398.1
XP_016876887.1
10
1157
XM_017021399.1
XP_016876888.1
9
1104
XM_017021400.1
XP_016876889.1
9
1022
XM_017021401.1
XP_016876890.1
10
1117
XM_017021402.1
XP_016876891.1
10
1028
XM_017021403.1
XP_016876892.1
9
1046
XM_017021404.1
XP_016876893.1
9
996
XM_017021405.1
XP_016876894.1
9
1063
XM_017021406.1
XP_016876895.1
10
1535
XM_017021407.1
XP_016876896.1
10
1044
XM_017021408.1
XP_016876897.1
8
980
XM_017021409.1
XP_016876898.1
9
1481
XM_017021410.1
XP_016876899.1
8
1002
XM_017021411.1
XP_016876900.1
8
1433
XM_017021414.1
XP_016876903.1
9
969
XM_017021415.1
XP_016876904.1
8
950
XM_017021416.1
XP_016876905.1
7
845
XM_017021417.1
XP_016876906.1
8
902
XM_017021418.1
XP_016876907.1
7
900
XM_017021419.1
XP_016876908.1
6
791
XM_017021420.1
XP_016876909.1
7
1320
GenBank mRNA
AF087905.1
-
9
1023
AF275678.1
-
10
1075
AK001344.1
-
8
968
AK225792.1
-
10
1082
AK303668.1
-
8
921
AK308055.1
-
9
1035
AK315620.1
-
9
1034
BC002566.2
-
9
1050
3138909
BC024208.2
-
9
1009
2960467
BC106012.1
-
7
794
BX161440.1
-
10
1053
KU178687.1
-
8
873
KU178688.1
-
9
921
KU178689.1
-
7
819
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
mRNA processing
nucleotide binding
RNA binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-8tsm6:80/100.66.77.47:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline