Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1585 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001109.1 | NP_001001109.1 | 1 | 622 |
Gene Symbol: LOC102550614 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250349.2 | XP_006250411.1 | 2 | 442 |