Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADIG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018082.2 | NP_001018092.1 | 1 | 155 | ||
NR_130907.1 | 1 | 155 | ||||
GenBank mRNA | BC029594.1 | - | 1 | 155 | 5273249 | |
BC119704.2 | - | 1 | 149 | 40117136 | ||
BC119705.2 | - | 1 | 149 | 40117137 |