Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163485.1 | NP_001156957.1 | 3 | 168 | ||
NM_009083.4 | NP_033109.1 | 2 | 224 | |||
GenBank mRNA | AK002988.1 | - | 2 | 222 | ||
AK137031.1 | - | 3 | 115 | |||
BC002060.1 | - | 2 | 138 | 3489311 | ||
BC069949.1 | - | 2 | 159 | |||
BC086890.1 | - | 3 | 118 | 6763356 | ||
BC086891.1 | - | 3 | 114 | 30122396 | ||
BC092137.1 | - | 2 | 85 | 6823165 | ||
BC100608.1 | - | 3 | 117 | 6753186 | ||
BG370645.1 | - | 3 | 168 | |||
CJ048578.1 | - | 2 | 225 | |||
CO806526.1 | - | 3 | 136 |
Gene Symbol: Gm12191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BG312195.1 | - | 1 | 48 |