Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009112.2 | NP_033138.1 | 2 | 153 | ||
GenBank mRNA | AK149077.1 | - | 2 | 139 | ||
AK161647.1 | - | 2 | 137 | |||
AK170919.1 | - | 2 | 133 | |||
BC025044.1 | - | 2 | 84 | 4949465 | ||
BU559648.1 | - | 156 | ||||
M16465.1 | - | 2 | 106 |