Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009267.2 | NP_033293.2 | 3, 4 | 442 | ||
GenBank mRNA | AK006124.1 | - | 3, 4 | 440 | ||
BC030164.1 | - | 3, 4 | 435 | |||
BC106151.1 | - | 3, 4 | 426 | 4923478 | ||
CK616968.1 | - | 3, 4 | 443 | |||
M33974.1 | - | 3, 4 | 438 | |||
X00349.1 | - | 3, 4 | 439 |