Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubb4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009451.3 | NP_033477.2 | 4 | 1912 | ||
GenBank mRNA | AK013717.1 | - | 4 | 1730 | ||
BC049112.1 | - | 4 | 1912 | 6415577 | ||
BC054831.1 | - | 4 | 1711 | 6306945 |