Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009741.4 | NP_033871.2 | 1 | 214 | ||
NM_177410.2 | NP_803129.2 | 1 | 214 | |||
GenBank mRNA | AK049473.1 | - | 1 | 209 | ||
CJ161448.1 | - | 1 | 215 |