Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018132.3 | NP_060602.2 | 6 | 557 | ||
XM_005249205.1 | XP_005249262.1 | 6 | 618 | |||
XM_011514723.2 | XP_011513025.2 | 5 | 493 | |||
GenBank mRNA | AK001407.1 | - | 1 | 107 | ||
AK293016.1 | - | 6 | 562 | |||
BC016279.1 | - | 6 | 547 | 3908895 |