Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079869.1 | NP_001073338.1 | 3 | 636 | ||
NM_010458.2 | NP_034588.2 | 1 | 559 | |||
GenBank mRNA | S35628.1 | - | 1 | 559 | ||
S35738.1 | - | 3 | 637 | |||
X66177.1 | - | 1 | 559 |