Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC100688.1 | - | 1 | 50 | ||
BC100689.1 | - | 1 | 51 | |||
BC100698.1 | - | 1 | 51 | |||
BC100699.1 | - | 1 | 51 | |||
BC114423.1 | - | 1 | 45 |