Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oxt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011025.4 | NP_035155.1 | 3 | 444 | ||
GenBank mRNA | AK003042.1 | - | 3 | 441 | ||
BC117030.1 | - | 3 | 423 | 40126349 | ||
BC117032.1 | - | 3 | 423 | 40126351 |