Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5orf45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017987.2 | NP_001017987.1 | 2 | 149 | ||
NM_016175.3 | NP_057259.2 | 2 | 149 | |||
GenBank mRNA | AK293901.1 | - | 2 | 115 | ||
AK295104.1 | - | 2 | 142 | |||
AK298954.1 | - | 2 | 106 | |||
BC056889.1 | - | 2 | 102 | |||
BC069051.1 | - | 2 | 117 | 3945859 | ||
BF965313.1 | - | 2 | 150 | |||
BQ642639.1 | - | 2 | 105 | |||
BX537968.1 | - | 2 | 170 | |||
BX647375.1 | - | 2 | 117 | |||
CN363239.1 | - | 2 | 73 | |||
CR936785.1 | - | 2 | 170 |