Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5orf45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017987.2 | NP_001017987.1 | 2 | 141 | ||
NM_016175.3 | NP_057259.2 | 2 | 141 | |||
GenBank mRNA | AK293901.1 | - | 2 | 107 | ||
AK295104.1 | - | 2 | 134 | |||
AK298954.1 | - | 2 | 98 | |||
BC056889.1 | - | 2 | 94 | |||
BC069051.1 | - | 2 | 109 | 3945859 | ||
BF965313.1 | - | 2 | 142 | |||
BQ642639.1 | - | 2 | 97 | |||
BX537968.1 | - | 2 | 162 | |||
BX647375.1 | - | 2 | 109 | |||
CN363239.1 | - | 2 | 65 | |||
CR936785.1 | - | 2 | 162 |