Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn1r132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122682.1 | NP_001116154.1 | 1 | 2344 | ||
GenBank mRNA | BC099438.1 | - | 709 |
Gene Symbol: Vmn1r148 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030736.2 | NP_109661.1 | 1 | 2379 | ||
GenBank mRNA | AF324872.1 | - | 1 | 2381 |