Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OVCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080822.2 | NP_543012.1 | 1 | 187 | ||
GenBank mRNA | AF073528.1 | - | 1 | 122 | ||
AF321875.1 | - | 1 | 122 | |||
BC040696.2 | - | 1 | 137 | 6158258 | ||
BC041170.1 | - | 1 | 137 | 5738727 | ||
CB137429.1 | - | 1 | 187 |
Gene Symbol: DPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK098268.1 | - | 2 | 1353 |