Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn1r214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134214.1 | NP_598975.1 | 1 | 352 | ||
GenBank mRNA | BC145835.1 | - | 1 | 396 | 40131138 |