Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018154.2 | NP_060624.1 | 2 | 333 | ||
GenBank mRNA | AB104486.1 | - | 2 | 161 | ||
AF279307.1 | - | 2 | 254 | |||
AK001288.1 | - | 2 | 292 | |||
AK001466.1 | - | 2 | 324 | |||
AK223080.1 | - | 2 | 287 | |||
AK302134.1 | - | 2 | 294 | |||
BC007726.2 | - | 2 | 257 | 4303318 | ||
BC010014.2 | - | 2 | 267 | 4304756 | ||
BC036521.1 | - | 2 | 297 | 5270216 | ||
CN426445.1 | - | 2 | 668 | |||
CR457235.1 | - | 2 | 161 |