Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr620 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146812.2 | NP_667023.2 | 1 | 628 | ||
GenBank mRNA | BC106798.1 | - | 1 | 628 | 40042289 | |
BC106799.1 | - | 1 | 628 | 40042291 |