Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146967.1 | NP_667178.1 | 1 | 910 | ||
GenBank mRNA | BC106781.1 | - | 1 | 910 | 40040933 | |
BC106782.1 | - | 1 | 910 | 40040934 |