Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr658 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147049.4 | NP_667260.1 | 2 | 1197 | ||
GenBank mRNA | AK033992.1 | - | 2 | 1198 | ||
BC104120.2 | - | 1 | 776 | 40044139 | ||
BC104121.1 | - | 1 | 776 |