Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153068.3 | NP_694708.2 | 2 | 558 | ||
GenBank mRNA | AK029240.1 | - | 2 | 502 | ||
AK031203.1 | - | 2 | 560 | |||
AK046566.1 | - | 2 | 500 | |||
AK048356.1 | - | 2 | 503 | |||
AK050243.1 | - | 2 | 538 | |||
AK084455.1 | - | 2 | 501 | |||
AK085016.1 | - | 2 | 501 | |||
AY531389.1 | - | 2 | 497 | |||
BC027084.1 | - | 3 | 205 | 5356224 | ||
BC031731.1 | - | 423 | ||||
BC113161.1 | - | 3 | 516 | 40060184 |