Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291508.1 | NP_001278437.1 | 7 | 1728 | ||
NM_001291509.1 | NP_001278438.1 | 6 | 1568 | |||
NM_008783.3 | NP_032809.1 | 7 | 1728 | |||
NM_183355.3 | NP_899198.1 | 8 | 1841 | |||
GenBank mRNA | AF020196.1 | - | 8 | 1532 | ||
AK087587.1 | - | 6 | 852 | |||
BC002244.1 | - | 7 | 954 | 3492658 | ||
BC054376.1 | - | 7 | 1689 | |||
BC058390.1 | - | 8 | 1569 | 5701148 | ||
HM369110.1 | - | 6 | 903 | |||
L27453.1 | - | 7 | 1107 |