Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018176.3 | NP_060646.2 | 1 | 239 | ||
XM_011513850.2 | XP_011512152.1 | 1 | 141 | |||
XM_017008356.1 | XP_016863845.1 | 1 | 141 | |||
GenBank mRNA | AB067503.1 | - | 1 | 132 | ||
AF467955.1 | - | 1 | 140 | |||
AJ487516.1 | - | 1 | 140 | |||
AJ487958.1 | - | 1 | 162 | |||
BC101759.1 | - | 1 | 239 | 8069265 | ||
BC101761.1 | - | 1 | 239 | 8069267 |