Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PERM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291366.1 | NP_001278295.1 | 4 | 3397 | ||
NM_001291367.1 | NP_001278296.1 | 5 | 3044 | |||
XM_017002583.1 | XP_016858072.1 | 4 | 3499 | |||
XM_017002584.1 | XP_016858073.1 | 4 | 3522 | |||
XM_017002585.1 | XP_016858074.1 | 3 | 3888 | |||
GenBank mRNA | AK123855.1 | - | 5 | 3020 | ||
BC006300.2 | - | 2 | 1194 | |||
F22110.1 | - | 1 | 203 |