Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAND2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003679.2 | 3 | 326 | |||
NR_136192.1 | 3 | 320 | ||||
NR_136193.1 | 3 | 317 | ||||
NR_136194.1 | 3 | 487 | ||||
NR_136195.1 | 3 | 320 | ||||
NR_136197.1 | 3 | 391 | ||||
NR_136198.1 | 2 | 208 | ||||
NR_136199.1 | 4 | 530 | ||||
NR_136200.1 | 3 | 320 | ||||
NR_136201.1 | 3 | 487 | ||||
GenBank mRNA | AB075501.1 | - | 3 | 229 | ||
AK123668.1 | - | 3 | 1383 | |||
AK300846.1 | - | 3 | 398 | |||
AU253586.1 | - | 3 | 229 | |||
BC101437.1 | - | 1 | 40 | |||
BC101981.1 | - | 1 | 40 | |||
BG519698.1 | - | 2 | 87 | |||
BQ882603.1 | - | 3 | 320 | |||
DA665500.1 | - | 3 | 396 | |||
DA863153.1 | - | 2 | 194 | |||
DC386781.1 | - | 4 | 441 | |||
DC421261.1 | - | 3 | 391 | |||
EF529495.1 | - | 3 | 326 | |||
EF529496.1 | - | 3 | 326 | |||
EF529497.1 | - | 3 | 326 | |||
EF529498.1 | - | 3 | 326 | |||
EF529499.1 | - | 3 | 233 | |||
EF529500.1 | - | 3 | 326 | |||
EF529501.1 | - | 2 | 121 | |||
EF529502.1 | - | 2 | 121 | |||
EF529503.1 | - | 3 | 400 | |||
EF529504.1 | - | 4 | 442 | |||
EF529505.1 | - | 3 | 326 | |||
EF529506.1 | - | 3 | 326 | |||
EF529507.1 | - | 3 | 326 | |||
EF529508.1 | - | 3 | 326 | |||
EF529509.1 | - | 3 | 326 | |||
EU000853.1 | - | 3 | 233 | |||
EU000854.1 | - | 3 | 233 | |||
EU000855.1 | - | 3 | 233 | |||
EU000856.1 | - | 3 | 233 |