Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAND2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136192.1 | 4 | 984 | |||
NR_136193.1 | 4 | 1681 | ||||
NR_136194.1 | 4 | 1151 | ||||
NR_136197.1 | 4 | 1055 | ||||
NR_136198.1 | 3 | 872 | ||||
NR_136199.1 | 5 | 1194 | ||||
NR_136200.1 | 4 | 1684 | ||||
NR_136201.1 | 4 | 1851 | ||||
GenBank mRNA | AB075501.1 | - | 4 | 1592 | ||
AK021601.1 | - | 1 | 1331 | |||
AK300846.1 | - | 4 | 1062 | |||
BG519698.1 | - | 3 | 749 | |||
EU000853.1 | - | 4 | 897 | |||
EU000854.1 | - | 4 | 897 | |||
EU000855.1 | - | 3 | 1989 | |||
EU000856.1 | - | 3 | 1989 |