Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAND2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136192.1 | 4 | 1379 | |||
NR_136193.1 | 4 | 2076 | ||||
NR_136194.1 | 4 | 1546 | ||||
NR_136197.1 | 4 | 1450 | ||||
NR_136198.1 | 3 | 1267 | ||||
NR_136199.1 | 5 | 1589 | ||||
NR_136200.1 | 4 | 2079 | ||||
NR_136201.1 | 4 | 2246 | ||||
GenBank mRNA | AB075501.1 | - | 4 | 1987 | ||
AK021601.1 | - | 1 | 1726 | |||
DQ831955.1 | - | 1 | 139 | |||
DQ831956.1 | - | 1 | 139 | |||
EF529498.1 | - | 4 | 514 | |||
EU000853.1 | - | 4 | 1292 | |||
EU000854.1 | - | 4 | 1292 | |||
EU000855.1 | - | 3 | 2384 | |||
EU000856.1 | - | 3 | 2384 |