Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012401.3 | NP_036533.2 | 28 | 4721 | ||
XM_005261909.1 | XP_005261966.1 | 29 | 4769 | |||
XM_005261910.2 | XP_005261967.1 | 29 | 4780 | |||
XM_005261911.4 | XP_005261968.1 | 28 | 4719 | |||
XM_006724413.2 | XP_006724476.1 | 30 | 4839 | |||
XM_011530682.2 | XP_011528984.1 | 29 | 4920 | |||
XM_017028703.1 | XP_016884192.1 | 30 | 4820 | |||
XM_017028704.1 | XP_016884193.1 | 29 | 4773 | |||
GenBank mRNA | AB002313.1 | - | 27 | 4638 | ||
AB075511.1 | - | 13 | 1831 | |||
AK025415.1 | - | 3 | 308 | |||
AK025701.1 | - | 15 | 2089 | |||
AK056543.1 | - | 11 | 1890 | |||
AK074932.1 | - | 10 | 1357 | |||
AK123131.1 | - | 1 | 226 | |||
BC004542.2 | - | 5 | 540 | |||
BT006887.1 | - | 5 | 524 | |||
S76730.1 | - | 6 | 871 |