Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC146880 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026899.1 | 11 | 2354 | |||
GenBank mRNA | AK055342.1 | - | 10 | 2225 |