Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TESPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098815.2 | NP_001092285.1 | 9 | 1386 | ||
NM_001136030.2 | NP_001129502.1 | 9 | 1483 | |||
NM_001261844.1 | NP_001248773.1 | 8 | 1235 | |||
NM_014796.2 | NP_055611.1 | 7 | 1326 | |||
XM_005269247.2 | XP_005269304.1 | 8 | 1309 | |||
XM_006719715.3 | XP_006719778.1 | 11 | 1817 | |||
XM_011539035.2 | XP_011537337.1 | 11 | 1830 | |||
XM_011539036.1 | XP_011537338.1 | 10 | 1651 | |||
XM_011539037.2 | XP_011537339.1 | 9 | 5871 | |||
XM_017020262.1 | XP_016875751.1 | 11 | 2880 | |||
XM_017020263.1 | XP_016875752.1 | 11 | 2786 | |||
GenBank mRNA | AB018291.1 | - | 7 | 1524 | ||
AF161375.1 | - | 1 | 232 | |||
AK295026.1 | - | 7 | 1130 | |||
AK298405.1 | - | 7 | 1152 | |||
AK303223.1 | - | 9 | 1409 | |||
AK315972.1 | - | 8 | 1189 | |||
BC044578.1 | - | 7 | 1156 | 5275264 | ||
BC132732.1 | - | 9 | 1386 | 40146754 | ||
BC132734.1 | - | 9 | 1386 | 40146756 |