Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00889 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026935.1 | 1 | 2846 | |||
GenBank mRNA | AK055234.1 | - | 1 | 2395 | ||
AK055382.1 | - | 1 | 2530 |