Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00889 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026935.1 | 1 | 1578 | |||
GenBank mRNA | AK055234.1 | - | 1 | 1127 | ||
AK055382.1 | - | 1 | 1262 | |||
BC105719.1 | - | 1 | 1252 | |||
BC105725.1 | - | 1 | 1252 | |||
BC105726.1 | - | 1 | 1252 | |||
BC105727.1 | - | 1 | 1252 | |||
BC139831.1 | - | 1 | 1255 |