Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA31D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207416.2 | NP_997299.2 | 4 | 4191 | ||
GenBank mRNA | AK126070.1 | - | 1 | 2895 |