Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080449.2 | NP_001073918.2 | 3 | 500 | ||
NR_102264.1 | 4 | 480 | ||||
XM_006717680.3 | XP_006717743.1 | 4 | 880 | |||
XM_017015799.1 | XP_016871288.1 | 1 | 35 | |||
GenBank mRNA | AK308333.1 | - | 3 | 500 | ||
AK308358.1 | - | 1 | 141 | |||
BC063664.1 | - | 3 | 413 | |||
BC111740.1 | - | 4 | 480 | 40038149 | ||
D42046.1 | - | 3 | 442 | |||
DC404138.1 | - | 3 | 501 |