Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015306.2 | NP_056121.2 | 47 | 5721 | ||
XM_005270690.3 | XP_005270747.1 | 34 | 3762 | |||
XM_017000831.1 | XP_016856320.1 | 48 | 5786 | |||
XM_017000832.1 | XP_016856321.1 | 48 | 5783 | |||
XM_017000833.1 | XP_016856322.1 | 48 | 5278 | |||
XM_017000834.1 | XP_016856323.1 | 47 | 5218 | |||
GenBank mRNA | AB028980.1 | - | 6 | 540 | ||
AK127075.1 | - | 30 | 3466 | |||
AL832370.1 | - | 9 | 947 |