Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABGGTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | Y08201.1 | - | 1191 |
Gene Symbol: PCLO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014510.2 | NP_055325.2 | 2 | 948 | ||
NM_033026.5 | NP_149015.2 | 2 | 948 | |||
XM_017012006.1 | XP_016867495.1 | 2 | 2223 | |||
XM_017012007.1 | XP_016867496.1 | 2 | 2207 | |||
GenBank mRNA | AK297315.1 | - | 2 | 951 | ||
BC122565.1 | - | 2 | 599 | |||
BC125271.1 | - | 2 | 599 | |||
Y19188.1 | - | 2 | 551 |