Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFWD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018124.3 | NP_060594.3 | 7, 8 | 1290 | ||
XM_005256021.4 | XP_005256078.1 | 8, 9 | 1423 | |||
XM_005256022.4 | XP_005256079.1 | 7, 8 | 2173 | |||
XM_006721228.3 | XP_006721291.1 | 8, 9 | 1423 | |||
XM_011523191.2 | XP_011521493.1 | 7, 8 | 1243 | |||
XM_017023391.1 | XP_016878880.1 | 8, 9 | 1339 | |||
XM_017023392.1 | XP_016878881.1 | 7, 8 | 1287 | |||
GenBank mRNA | AK001382.1 | - | 2, 3 | 192 | ||
AK291200.1 | - | 8, 9 | 1390 | |||
AK315786.1 | - | 4, 5 | 434 | |||
BC002574.2 | - | 5, 6 | 667 | |||
BC059371.1 | - | 7, 8 | 1239 | 5266451 |