Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYSM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085487.2 | NP_001078956.1 | 17 | 2078 | ||
XM_006710314.3 | XP_006710377.1 | 17 | 2094 | |||
XM_011540573.2 | XP_011538875.1 | 18 | 2058 | |||
XM_011540574.2 | XP_011538876.1 | 17 | 2326 | |||
XM_017000195.1 | XP_016855684.1 | 18 | 2130 | |||
XM_017000196.1 | XP_016855685.1 | 15 | 1980 | |||
XM_017000197.1 | XP_016855686.1 | 13 | 1758 | |||
GenBank mRNA | AB067502.1 | - | 13 | 2104 | ||
AK126835.1 | - | 11 | 1627 | |||
AK292919.1 | - | 4 | 765 | |||
BX537912.1 | - | 4 | 342 | |||
BX648168.1 | - | 14 | 1900 |