Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175732.2 | NP_783859.1 | 3, 4 | 634 | ||
GenBank mRNA | BC014048.1 | - | 2, 3 | 280 | ||
BC020242.1 | - | 3, 4 | 463 | 4591438 | ||
BQ050653.1 | - | 1, 2 | 27 | |||
DA476984.1 | - | 3, 4 | 490 | |||
DR004030.1 | - | 3, 4 | 492 |